
5-(4-Nitrophenyl)-1H-tetrazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16687-60-8
:
Formula:C₇H₅N₅O₂
MDL Number:MFCD00068729
:191.15 g/mol
Melting Pt:∼ 223 °C (dec.)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together