Order Entry
Poland
ContactUsLinkComponent
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid 97%
Corrosive
Irritant
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid 97%
Catalog # L09516.06
CAS Number:  
Corrosive
Irritant
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid 97%
Catalog # L09516.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :226.08 g/mol
Melting Pt:85…89 °C
Storage Temperature:Ambient
ADR:8,III
 :GHS05
 :GHS07
CAS Number:1547-36-0
Formula:C₅H₄F₆O₃
MDL Number:MFCD00040910