
7-Ethoxy-7-oxoheptanoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:33018-91-6
:MFCD00801202
Formula:C₉H₁₆O₄
MDL Number:MFCD00014383
:188.22 g/mol
Boiling Pt:135…138 °C (1 mmHg)
Melting Pt:10 °C
Density:1,06
Flash Pt:>110 °C (230 °F)
EINECS:251-346-6
:GHS07
Storage Temperature:Temperatura otoczenia
Related Information
Frequently Bought Together