Cycloheximide 95%, Ultrapure
1 g
Supplier: Thermo Fisher Scientific
Cycloheximide 95%, Ultrapure
Catalog # J66901.03
Supplier: Thermo Fisher Scientific
CAS Number: 66-81-9
Jump to:
Product Details & Documents
Applications: A protein synthesis inhibitor in eukaryotic cells.
Applications: A protein synthesis inhibitor in eukaryotic cells
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Number:66-81-9
- Formula:C₁₅H₂₃NO₄
- MDL Number:MFCD00082346
- Molecular Weight:281.35 g/mol
- Merck Index:14,2728
- Boiling Pt:492 °C
- Melting Pt:111…116 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:200-636-0
- UN:2811
- ADR:6.1,I
Documents
Specifications
Specifications
- Pk:1 g



