Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
J65608.03
:
Thermo Fisher Scientific
:
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
J65608.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
:
355.32 g/mol
:
Refrigerator
:
C₁₄H₁₈FN₅O₅
:
MFCD15145246