Order Entry
Poland
ContactUsLinkComponent
5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine 98%
5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine 98%
Catalog # J65583.03
CAS Number:  
5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine 98%
Catalog # J65583.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :669.72 g/mol
CAS Number:114745-26-5
Formula:C₃₆H₃₉N₅O₈
MDL Number:MFCD09842119