Order Entry
Poland
ContactUsLinkComponent
2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
Irritant
2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
Catalog # J64938.MB
CAS Number:  
Irritant
2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
Catalog # J64938.MB
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :1272.511 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:95648-78-5
Formula:C₃₀H₄₅N₆Na₃O₃₃P₆
MDL Number:MFCD00084748