Order Entry
Poland
ContactUsLinkComponent
 
TRIS-buffered saline (TBS, 10X) pH 8,0
 
TRIS-buffered saline (TBS, 10X) pH 8,0
 

 

  • CAS No.:
    77-86-1
  • MDL No.:
    MFCD00004679
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

 

Frequently Bought Together