
Alendronate sodium trihydrate 97%
Catalog # J61397.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
CAS Number:121268-17-5
:
Formula:C₄H₁₂NaNO₇P₂·3H₂O
MDL Number:MFCD01748233
Merck Index:14,229
:325.12 g/mol
:GHS07