Order Entry
Poland
ContactUsLinkComponent
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride 99%
Irritant
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride 99%
Catalog # J61105.03
CAS Number:  
Irritant
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride 99%
Catalog # J61105.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

 :
 :276.20 g/mol
Merck Index:14,1499
Melting Pt:233…234 °C
EINECS:250-759-9
 :GHS07
Storage Temperature:Ambient
CAS Number:31677-93-7
Formula:C₁₃H₁₉Cl₂NO
MDL Number:MFCD00055209