Order Entry
Poland
ContactUsLinkComponent
 
Boc-Phe-Phe-OH
 
Boc-Phe-Phe-OH
 

 

  • CAS No.:
    13122-90-2
  • Storage conditions:
    Ambient
  • MDL No.:
    MFCD00190824
  • Protein synonyms:
    N-Boc-L-phenylalanyl-L-phenylalanine
  • Protein/peptide name:
    Boc-Phe-Phe-OH
  • Purity:
    94,0%
  • Molecular weight:
    412,486 g/mol
  • Molecular formula:
    C₂₃H₂₈N₂O₅

 

Frequently Bought Together