
1-Oxa-6-azaspiro[3.4]octane oxalate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:C₆H₁₁NO·C₂H₂O₄
MDL Number:MFCD18325163
:203.19 g/mol
:GHS07
Storage Temperature:Zamra·arka
Related Information
Frequently Bought Together