(4-Bromophenylethynyl)trimethylsilane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16116-78-2
:
Formula:C₁₁H₁₃BrSi
MDL Number:MFCD01863556
:253.214 g/mol
Melting Pt:61…63 °C
Storage Temperature:Ambient
Frequently Bought Together