

Cyclohexylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:931-51-1
:
Formula:C₆H₁₁ClMg
MDL Number:MFCD00003816
:142.91 g/mol
UN:2924
ADR:3,II
:GHS02
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together