5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2251-79-8
:MFCD00052525
Formula:C₈H₅F₃N₄
MDL Number:MFCD06248886
:214.15 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together