Order Entry
Poland
ContactUsLinkComponent
4-Chloro-5-phenylthieno[2,3-d]pyrimidine 98%
Irritant
4-Chloro-5-phenylthieno[2,3-d]pyrimidine 98%
Catalog # H50537.03
CAS Number:  
Irritant
4-Chloro-5-phenylthieno[2,3-d]pyrimidine 98%
Catalog # H50537.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:182198-35-2
 :
Formula:C₁₂H₇ClN₂S
MDL Number:MFCD00218029
 :246.72 g/mol
Melting Pt:125…129 °C
 :GHS07
Storage Temperature:Ambient