3-(4-Methoxyphenyl)propionyl chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15893-42-2
:
Formula:C₁₀H₁₁ClO₂
MDL Number:MFCD02258671
:198.65 g/mol
ADR:8,II
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together