
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:368-63-8
:
Formula:C₁₀H₈F₆O
MDL Number:MFCD01632521
:258.16 g/mol
Melting Pt:70…74 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together