Order Entry
Poland
ContactUsLinkComponent
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 95%
Irritant
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 95%
Catalog # H31692.MD
CAS Number:  
Irritant
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 95%
Catalog # H31692.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:1315281-29-8
 :
Formula:C₁₀H₁₆BClN₂O₂
MDL Number:MFCD11100959
 :242.52 g/mol
Melting Pt:131…135 °C
 :GHS07