
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:374-25-4
:
Formula:C₄H₃BrClF₃
MDL Number:MFCD00039225
:223.43 g/mol
Boiling Pt:98…99 °C
Density:1.678
:GHS07
Storage Temperature:Ambient
Frequently Bought Together