Farnesyl acetate (mixture of isomers) mixture of isomers 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29548-30-9
:
Formula:C₁₇H₂₈O₂
MDL Number:MFCD00036516
:264.41 g/mol
Density:0.91
Flash Pt:95 °C (203 °F)
Storage Temperature:Ambient
EINECS:249-689-1
Related Information
Frequently Bought Together