4-(2-Furyl)-3-buten-2-one cis- and trans-mixture 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-(Furan-2-yl)but-3-en-2-one, Furfuralacetone
CAS Number:623-15-4
:
Formula:C₈H₈O₂
MDL Number:MFCD00039566
Boiling Pt:112…115 °C (10 mmHg)
Molecular Weight:136.15 g/mol
Melting Pt:33…38 °C
Flash Pt:103 °C (217 °F)
EINECS:210-774-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together