
Iron(II,III) oxide 97%, nanopowder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1317-61-9
:
Formula:Fe₃O₄
MDL Number:MFCD00011010
:231.54 g/mol
Merck Index:14,4040
Melting Pt:1538 °C
Density:5.2
EINECS:215-277-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together