Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium oxide
Aluminium oxide, Drysphere®, without indicator
Aluminium oxide, Drysphere®, without indicator
46172.22
:
Thermo Fisher Scientific
:
Aluminium oxide, Drysphere®, without indicator
46172.22
:
Thermo Fisher Scientific
:
Pk:
100 g
Seq:
3401
:
:
101.96 g/mol
:
14,356
:
2980 °C
:
2045 °C
:
Ambient
:
215-691-6
:
1344-28-1
:
11092-32-3
:
Al₂O₃
:
MFCD00003424