Order Entry
Poland
ContactUsLinkComponent
Palladium 5% on activated carbon paste (50% water), sulfided A103038-5
Palladium 5% on activated carbon paste (50% water), sulfided A103038-5
Catalog # 45497.09
CAS Number:  
Palladium 5% on activated carbon paste (50% water), sulfided A103038-5
Catalog # 45497.09
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

CAS Number:7440-05-3
 :
Formula:Pd
MDL Number:MFCD03457879
 :106.420 g/mol
Storage Temperature:Ambient
EINECS:231-115-6
MDL Number:MFCD00011167