Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9016-00-6
:
Formula:(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
MDL Number:MFCD00148360
Density:0.974
Storage Temperature:Ambient
Related Information
Frequently Bought Together