Hafnium(IV) sulphate ≥99.9% (metals basis excluding Zr) (max. 1% Zr)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15823-43-5
:
Formula:Hf(SO₄)₂
MDL Number:MFCD00168081
:370.62 g/mol (anhy)
Storage Temperature:Ambient
Related Information
Frequently Bought Together