Zirconium oxide catalyst support
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12036-01-0
:
Formula:OZr
MDL Number:MFCD00011310
:107.22 g/mol
Storage Temperature:Ambient
EINECS:215-227-2
Related Information
Frequently Bought Together