

Ruthenium(IV) sulphide ≥99.9% (metals basis)
Catalog # 43098.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:4.1,III
:165.19 g/mol
EINECS:235-318-0
UN:3178
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:12166-20-0
Formula:RuS₂
MDL Number:MFCD00799846