
Aluminum dihydrogen phosphate 50% (w/w) in aqueous solution
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Sodium-2-naphthalenesulphonate 98% may cont. up to 10% residual inorganic salts and water
: Thermo Fisher Scientific
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals