Order Entry
Poland
ContactUsLinkComponent
Cobalt(II)-2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99% (metals basis)
Health
Flammable
Irritant
Cobalt(II)-2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99% (metals basis)
Catalog # 42819.AC
CAS Number:  
Health
Flammable
Irritant
Cobalt(II)-2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99% (metals basis)
Catalog # 42819.AC
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

About this item

 :
 :209.11 g/mol
ADR:3,II
 :Packaged under argon in resealable ChemSeal™ bottles.
UN:1188
 :GHS08
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:142600-61-1
Formula:C₆H₁₄CoO₄
MDL Number:MFCD00798554