

Molybdenum(V) isopropoxide 5% (w/v) in isopropanol (IPA) , 99.6% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:209733-38-0
:
Formula:C₁₅H₃₅MoO₅
MDL Number:MFCD00210636
:391.39 g/mol
ADR:3,II
:Packaged under argon in resealable ChemSeal™ bottles.
Flash Pt:12 °C (54 °F)
UN:1219
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together