Order Entry
Poland
ContactUsLinkComponent
Zirconium(IV) oxide-2-ethylhexanoate (∼6% Zr) in mineral spirits
Flammable
Irritant
Zirconium(IV) oxide-2-ethylhexanoate (∼6% Zr) in mineral spirits
Catalog # 39543.36
CAS Number:  
Flammable
Irritant
Zirconium(IV) oxide-2-ethylhexanoate (∼6% Zr) in mineral spirits
Catalog # 39543.36
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
UN:1268
ADR:3,III
 :GHS02
 :GHS07
CAS Number:22464-99-9
Formula:(C₈H₁₅O₂)xZn
MDL Number:MFCD00072684