Palladium hydroxide on carbon ≥99% (Pearlman's catalyst)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12135-22-7
:
Formula:H₂O₂Pd
MDL Number:MFCD00064599
:140.435 g/mol
Storage Temperature:Ambient
EINECS:235-219-2
Related Information
Frequently Bought Together