Platinum 0.5% on carbon, granules, reduced
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-06-4
:
Formula:Pt
MDL Number:MFCD00011179
:195.08 g/mol
Storage Temperature:Ambient
UN:1325
ADR:4.1,III
:GHS02
Related Information
Frequently Bought Together