Platinum 1% on activated carbon wetted with ca. 50% water, powder, reduced
Catalog # 38312.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
Specifications
About this item
:
:195.08 g/mol
Storage Temperature:Ambient
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179