Anti-TAP2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9824
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
- Antigen symbol:TAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6891
- Antigen synonyms:PSF-2|D6S217E|PSF2|RING11|ABCB3|APT2|ABC18
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human