Anti-KCNA4 Rabbit Polyclonal Antibody
Catalog # BSBTPB9807
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:potassium channel, voltage gated shaker related subfamily A, member 4
- Antigen symbol:KCNA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3739
- Antigen synonyms:HPCN2|PCN2|KCNA4L|HBK4|KV1.4|KCNA8|HUKII|HK1
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: KCNA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human