Anti-TNFSF8 Rabbit Polyclonal Antibody
Catalog # BSBTRP1103
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 8
- Antigen symbol:TNFSF8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Gene ID:944
- Antigen synonyms:CD30LG|CD153|CD30L
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TNFSF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human