Anti-F1LVT0 Rabbit Polyclonal Antibody
Catalog # ABGEAP3879A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:F1LVT0
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Modification:phospho-Ser277
- Storage buffer:pH 7,4
- Molecular weight:159.796 kDa
- Sequence:KPTTELKBLLQEQSC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.42
- Shipping temperature:4 °C
- Immunogen:This rat CCNB3 Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S277 of rat CCNB3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: F1LVT0
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat