Safety ankle boots, lace-up, Concept M, Performance
: U-Power
Mid S3 safety safety footwear, anti-static, made from water repellent full grain leather and a hard wearing, slip resistant PU sole.
- Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
- Thinsulate for thermal insulation
- Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
- Stress-out tongue system allows easy instep and provides added comfort when walking
- Natural fit width 11 for additional comfort
- Airtoe protective aluminium toecap with breathable membrane (200 J)
- Light, flexible and non-metallic puncture resistant midsole “Save & Flex”covers 100% of the sole of the foot
Ideal for outdoor applications.
Colour: Black
: EN 20345:2011; S3, SRC
Frequently Bought Together