Screw caps for wash bottles, wide mouth, Azlon
Dostawca: SciLabware
Certyfikaty
Informacje o tym produkcie
PP, coloured
- Availablle in six colours
- One-piece closures that maintain their shape in use
- Closures feature precision jet-tip aperture for fine control
Caps for wide neck wash bottles with dip-tube and patented DripLok® vapour venting valve that prevents solvent drips.
Frequently Bought Together