
3-Methylflavone-8-carboxylic acid ≥98% for synthesis, Sigma-Aldrich®
Catalog # 8.41878.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Melting Point:235 - 240 °C
- Molar Mass:280.28 g/mol
- Hill Formula:C17H12O4
- HS Code:2932 99 00
- Storage temperature:15...25 °C
- Assay (HPLC, area%):≥98% (a/a)
- Melting range (lower value):≥235 °C
- Melting range (upper value):≤240 °C
- Identity (IR):passes test
Specification Test Results
About this item
:
:280.28 g/mol
:M-Clarity™ quality level = MQ200
EINECS:222-425-2
:GHS07
Storage Temperature:Ambient
CAS Number:3468-01-7
Formula:C₁₇H₁₂O₄
MDL Number:MFCD00040998