Order Entry
Poland
ContactUsLinkComponent
Anti-HA Mouse Monoclonal Antibody (Biotin) [clone: 16B12]
Anti-HA Mouse Monoclonal Antibody (Biotin) [clone: 16B12]
Catalog # AGRIAS152922
Supplier:  Agrisera
Anti-HA Mouse Monoclonal Antibody (Biotin) [clone: 16B12]
Catalog # AGRIAS152922
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HA
  • Clonality:
    Monoclonal
  • Clone:
    16B12
  • Conjugation:
    Biotin
  • Host:
    Mouse
  • Western blot:
    Yes
  • Immunogen:
    HA-tag: CYPYDVPDYASL
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to biotin. This is of advantage to shorten assay time by no need to use a secondary antibody to HA. ,Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to biotin. This is of advantage to shorten assay time by no need to use a secondary antibody to HA.

Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone: 16B12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: