Anti-GPXh Rabbit Polyclonal Antibody
Catalog # AGRIAS152882
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:GPXH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Immunogen:KLH-conjugated synthetic peptide derived from glutathione peroxidase protein sequence from Chlamydomonas reinhardtii, UniProt: O22448
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Glutathione peroxidase (GPXh) is an enzyme from glutathione peroxidase family.,Glutathione peroxidase (GPXh) is an enzyme from glutathione peroxidase family.
Type: Primary
Antigen: GPXH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rabbit, Rat