Anti-LhcSR1 Rabbit Polyclonal Antibody
AGRIAS142819
: Agrisera
- Antibody type:Primary
- Antigen symbol:LHCSR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Spinacia oleracea,Zea mays,A. thaliana,H. vulgare
- Western blot:Yes
- Immunogen:KLH-conjugated synthetic peptide derived from LHCSR1 protein sequence from Chlamydomonas reinhardtii, UniProt: P93664
- Size:50 µl
- Pk:50 µl
LHCSR1 (Stress-related chlorophyll a/b binding protein 1) plays a role in an efficient enery dissipation process, called non-photochemical quenching (NPQ), in Chlamydomonas reinhardtii. ,LHCSR1 (Stress-related chlorophyll a/b binding protein 1) plays a role in an efficient enery dissipation process, called non-photochemical quenching (NPQ), in Chlamydomonas reinhardtii.
Type: Primary
Antigen: LHCSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, Spinacia oleracea, Zea mays