Specifications
- Antibody type:Primary
- Antigen symbol:GID1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Lupinus luteus
- Western blot:Yes
- Immunogen:recombinant full-length Arabidopsis thaliana GID1c, UniProt: Q940G6 TAIR: At5g27320
- Size:50 µg
- Pk:50 µG
Specifications
About this item
GID1c (Gibberellin receptor GID1C) is a gibberellin (GA) receptor ortholog of the rice GA receptor OsGID1 which binds to GA and shows affinity to GA4 and interacts with DELLA proteins in vivo in the presence of GA4. Alternative names: AtCXE19, Carboxylesterase 19, GID1-like protein 3, Protein GA INSENSITIVE DWARF 1C.,GID1c (Gibberellin receptor GID1C) is a gibberellin (GA) receptor ortholog of the rice GA receptor OsGID1 which binds to GA and shows affinity to GA4 and interacts with DELLA proteins in vivo in the presence of GA4. Alternative names: AtCXE19, Carboxylesterase 19, GID1-like protein 3, Protein GA INSENSITIVE DWARF 1C.
Type: Primary
Antigen: GID1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Lupinus luteus