Anti-RALA Rabbit Polyclonal Antibody
BSBTPB9794
- Antibody type:Primary
- Antigen name:Ras-related protein Ral-A
- Antigen symbol:RALA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5898
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Ras-related protein Ral-A(RALA) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: RALA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat