Anti-Rab11A Rabbit Polyclonal Antibody
BSBTPB9789
- Antibody type:Primary
- Antigen name:RAB11A, member RAS oncogene family
- Antigen symbol:RAB11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8766
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Ras-related protein Rab-11A(RAB11A) detection. Tested with WB in Human.
Type: Primary
Antigen: Rab11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human