Anti-Actin Rabbit Polyclonal Antibody
Catalog # BSBTPB9751
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Actin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:58
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Actin, alpha skeletal muscle(ACTA1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: Actin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat